missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP 96345, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective non-peptide NK1 receptor antagonist. Attenuates substance P-induced salivary response and inhibits neurogenic inflammation in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 412.57 |
Percent Purity | >98% |
Chemical Name or Material | CP 96345 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'