missing translation for 'onlineSavingsMsg'
Learn More

CP 96345, Tocris Bioscience™

Product Code. 18054592
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NK1 antagonist

Potent and selective non-peptide NK1 receptor antagonist. Attenuates substance P-induced salivary response and inhibits neurogenic inflammation in vivo.

Chemical Identifiers

CAS 132746-60-2
Molecular Formula C28H32N2O
Molecular Weight (g/mol) 412.577
InChI Key FLNYLINBEZROPL-NSOVKSMOSA-N
Synonym unii-w22ila2i52, chembl16192, 2-diphenylmethyl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, 2s,3s-cis-2-diphenylmethyl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, 2s,3s-2-diphenylmethyl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, 2s,3s-n-2-methoxyphenyl methyl-2-diphenylmethyl-1-azabicyclo 2.2.2 octan-3-amine, 1-azabicyclo 2.2.2 octan-3-amine, 2-diphenylmethyl-n-2-methoxyphenyl methyl-, 2s-cis, 2s,3s-2-benzhydryl-n-2-methoxyphenyl methyl-1-azabicyclo 2.2.2 octan-3-amine, d03sxn, 2-benzhydryl-1-aza-bicyclo 2.2.2 oct-3-yl-2-methoxy-benzyl-amine
PubChem CID 104943
IUPAC Name (2S,3S)-2-benzhydryl-N-[(2-methoxyphenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine
SMILES COC1=CC=CC=C1CNC2C3CCN(C2C(C4=CC=CC=C4)C5=CC=CC=C5)CC3

Specifications

Quantity 50 mg
Formula Weight 412.57
Percent Purity >98%
Chemical Name or Material CP 96345
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'