Learn More
Cole-Parmer™ CPC™ NS4 Non-spill Panel Mount Coupling Insert with Compression Connection
Virtually eliminate any loss of product with this leak-free design
Brand:  Cole-Parmer™ 31605-13
Product Code. 15497403
  • £24.00 / Each
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Less than 0.10mL spillage.
  • One-hand operation.
  • Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
  • For a complete fitting, order one body and one insert.
  • All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifications
Specifications
Coupling Insert, Non-Spill, Panel Mount, 3/8 in. OD Tube
3/8 in.
Quick-Disconnect x Compression
ABS Plastic