missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15184-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can bind the close sequences genes.
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable), together with a carnitine-acylcarnitine translocase. CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
CPT1A | |
Polyclonal | |
Unconjugated | |
CPT1A | |
CPT I, CPT1, CPT1 L, CPT1A, CPTI L, L CPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12894, 1374, 25757 | |
-20°C | |
Liquid |
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32198, P50416, P97742 | |
Cpt1a | |
CPT1A Fusion Protein Ag7202 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |