missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine, anhydrous, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | ∼290°C (decomposition) |
Density | 1.33 |
Assay Percent Range | 98% |
Quantity | 25 g |
Beilstein | 907175 |
Merck Index | 14,2568 |
Solubility Information | Soluble in water. |
Formula Weight | 131.13 |
Percent Purity | 98% |
Chemical Name or Material | Creatine, anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'