Learn More
Creatine monohydrate, 99%, Thermo Scientific Chemicals
Creatine monohydrate, CAS # 6020-87-7, is the monohydrate form of creatine, an amino acid found primarily in skeletal muscle cells that is important for rapid energy replenishment.
£26.30 - £116.00
Chemical Identifiers
CAS | 6020-87-7 |
---|---|
Molecular Formula | C4H9N3O2·H2O |
Molecular Weight (g/mol) | 149.15 |
MDL Number | MFCD00071582 |
InChI Key | MEJYXFHCRXAUIL-UHFFFAOYSA-N |
Synonym | creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure |
PubChem CID | 80116 |
IUPAC Name | 2-[carbamimidoyl(methyl)amino]acetic acid;hydrate |
SMILES | CN(CC(=O)O)C(=N)N.O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387980
|
Thermo Scientific Acros
226790250 |
25 g | Glass bottle |
£26.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783421
|
Thermo Scientific Acros
226791000 |
100 g | Glass bottle |
£37.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209163
|
Thermo Scientific Acros
226795000 |
500 g | Glass bottle |
£116.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6020-87-7 | |
149.15 | |
MEJYXFHCRXAUIL-UHFFFAOYSA-N | |
80116 | |
CN(CC(=O)O)C(=N)N.O |
C4H9N3O2·H2O | |
MFCD00071582 | |
creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure | |
2-[carbamimidoyl(methyl)amino]acetic acid;hydrate |
Specifications
6020-87-7 | |
White to Yellow | |
12.6% max. (105°C) | |
98.5% min. (on dry substance) (HPLC) | |
H2NC(=NH)N(CH3)CH2CO2H·H2O | |
04, 363 | |
creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure | |
MEJYXFHCRXAUIL-UHFFFAOYSA-N | |
2-[carbamimidoyl(methyl)amino]acetic acid;hydrate | |
80116 | |
99% | |
Creatine monohydrate |
292.0°C | |
1% max. | |
Authentic | |
C4H9N3O2·H2O | |
MFCD00071582 | |
14, 2568 | |
(1% in water) Clear colorless to light yellow | |
CN(CC(=O)O)C(=N)N.O | |
149.15 | |
149.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only