missing translation for 'onlineSavingsMsg'
Learn More

Creatine phosphate disodium salt tetrahydrate, 99%, Thermo Scientific Chemicals

Product Code. 10616794
Click to view available options
Quantity:
1 g
2 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71519-72-7
Molecular Formula C4H8N3Na2O5P
Molecular Weight (g/mol) 255.08
MDL Number MFCD00150192
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
Synonym creatine phosphate sodium salt
PubChem CID 91884830
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Na2O3PNHC(NH)N(CH3)CH2COOH·4H2O
Quantity 2 g
Solubility Information (10% in water) Clear colorless solution
IUPAC Name 2-[methyl-[(E)-N'-phosphonocarbamimidoyl]amino]acetic acid;sodium
Formula Weight 327.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Creatine phosphate disodium salt tetrahydrate
Show More Show Less
Product Identifier
  • Creatine phosphate disodium salt tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'