missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatinine, 98%, Thermo Scientific Chemicals
£34.67 - £288.44
Chemical Identifiers
CAS | 60-27-5 |
---|---|
Molecular Formula | C4H7N3O |
Molecular Weight (g/mol) | 113.12 |
InChI Key | DDRJAANPRJIHGJ-UHFFFAOYSA-N |
Synonym | creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu |
PubChem CID | 588 |
ChEBI | CHEBI:16737 |
IUPAC Name | 2-amino-3-methyl-4H-imidazol-5-one |
SMILES | CN1CC(=O)N=C1N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10553401
|
Thermo Scientific Acros
228940250 |
25 g | Glass bottle |
£34.67
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10033160
|
Thermo Scientific Acros
228940500 |
50 g | Glass bottle |
£64.52
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11916261
|
Thermo Scientific Acros
228942500 |
250 g | Glass bottle |
£288.44
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-27-5 | |
113.12 | |
creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu | |
CHEBI:16737 | |
CN1CC(=O)N=C1N |
C4H7N3O | |
DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
588 | |
2-amino-3-methyl-4H-imidazol-5-one |
Specifications
60-27-5 | |
White | |
0.5% (105°C) max. | |
98% | |
24,245 | |
creatinine, 2-imino-1-methylimidazolidin-4-one, 1-methylglycocyamidine, 1-methylhydantoin-2-imide, creatinine,heated, 4h-imidazol-4-one, 2-amino-1,5-dihydro-1-methyl, 2-amino-1-methylimidazolin-4-one, creatinina, kreatinin, unii-ayi8ex34eu | |
DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
2-amino-3-methyl-4H-imidazol-5-one | |
588 | |
113.12 | |
Crystalline Powder |
255.0°C | |
290°C | |
Authentic | |
C4H7N3O | |
15,2557 | |
(5% in water) Clear colorless to slightly yellow | |
CN1CC(=O)N=C1N | |
113.12 | |
CHEBI:16737 | |
98% | |
Creatinine |
Safety and Handling
missing translation for 'einecsNumber' : 200-466-7
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title