Learn More
Cresol Purple, Pure, Solution 0.04%, pH Indicator, Fisher Chemical
Brand: Fisher Chemical C/7450L/05
Code : 27
Additional Details : CAS Number : 2303-01-7 Weight : 0.10000kg
Description
100mL, Amber glass bottle
Specifications
2309-07-1 | |
5871 | |
208.21 | |
100 mL |
C11H12O4 | |
AUJXJFHANFIVKH-GQCTYLIASA-N | |
Amber glass bottle | |
Liquid |