missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red

Product Code. 11483467
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11483467

Brand: Thermo Scientific Chemicals A17243.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol

Cresol red is commonly used with lead acetate for the detection of urease after PAGE. It is also used as an indicator with the following transition intervals: pH 1.8 (orange) to pH 2.0 (yellow); pH 7.0 (yellow) to pH 8.8 (violet). Cresol Red is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approx. 125 bp DNA. Cresol Red does not inhibit Taq polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresol red is commonly used with lead acetate for the detection of urease after PAGE. It is also used as an indicator with the following transition intervals: pH 1.8 (orange) to pH 2.0 (yellow); pH 7.0 (yellow) to pH 8.8 (violet). Cresol Red is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approx. 125 bp DNA. Cresol Red does not inhibit Taq polymerase.

Solubility
Soluble in water (slightly at 20°C), 50% ethanol (0.1%), dil. acids, and dil. alkalis. Insoluble in benzene.

Notes
Transition interval (acid): pH 1.8 (orange) to pH 2.0 (yellow); (alkaline) pH 7.0 (yellow) to pH 8.8 (violet)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1733-12-6
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005878
InChI Key OBRMNDMBJQTZHV-UHFFFAOYSA-N
Synonym o-Cresolsulfonephthalein
PubChem CID 73013
ChEBI CHEBI:86218
IUPAC Name 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
SMILES CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Melting Point >300°C
Beilstein 343399
Merck Index 14,2578
Solubility Information Soluble in water (slightly at 20°C),50% ethanol (0.1%),dil. acids,and dil. alkalis. Insoluble in benzene.
Formula Weight 382.44
Quantity 25 g
Chemical Name or Material Cresol Red

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.