missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JG Finneran Associates™ Crimp Caps
Use these caps with standard 11mm chromatography vials. JG Finneran™ Crimp Caps feature an open top for filling and retrieving samples with a syringe.
Brand: JG Finneran Associates™ 5140-11G
This item is not returnable.
View return policy
Description
- Made of the highest quality aluminum
- With PTFE seal
- Seals are available in a variety of colors for easy sample identification
Specifications
Rubber | |
PTFE, Rubber, Aluminum | |
1000 Pk. | |
11 mm |
Red, Green | |
Clear PTFE/red Rubber | |
Crimp Cap |