Learn More
Macherey-Nagel™ Crimp Closures N8 with Hole
Aluminum crimp N8 closures have a red rubber/PTFE septa material for routine analysis. All closures are automatically assembled under strict hygienic conditions. They are compitable with Dionex, Perkin Elmer and Thermo Scientific autosamplers.
Brand:  Macherey-Nagel™ 702025
Product Code. 15855421
  • £9.75 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Red rubber/PTFE septa
  • N8 Diameter
  • Ideal for GC and HPLC applications
Specifications
Specifications
Red-Orange/Colorless
Silver
Red Rubber/PTFE
Temp. Resistance -40°C up to 110°C/Analytical Purity - Medium/Medium Hard to Penetrate/Medium Resealability
1.0 mm (Septa)
Dionex, PerkinElmer, Thermo Scientific Autosamplers
Aluminum Crip Cap with Hole, Prefitted Septa
Aluminum
GC, HPLC
N8
Crimp Seal Vial