missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Crimp Neck Vial, Clear Glass

Flat bottom.

Brand:  Fisherbrand™ 20 09 0343

40.70 GBP valid until 2025-03-14
Use promo code "24142" to get your promotional price.


 View more versions of this product

Product Code. 11560535

  • £48.35 / Pack of 88
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Crimp Neck
  • Flat bottom
  • Glass
  • li>Clear
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Glass
Storage
Polypropylene Box
100 mL
94.5 mm
3rd Hydrolytic Class
No
51.6 mm
118.8 mL
Crimp
6000 μL
Flat Bottom
100
ND20
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Crimp Neck Vial, Clear Glass > flat bottom,100ml,94.5mm height

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.