missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (6G12), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-52398G
Additional Details : Weight : 0.00970kg
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CRISPR-Cas9 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bacteria | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
6G12 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
Recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2] | |
0.1 mL | |
Gene Editing, Gene Knockouts, Gene Therapy | |
This CRISPR-Cas9 antibody (6G12) - C-terminus is specific to Cas9 from Streptococcus pyogene. | |
Store at 4C in the dark. | |
IgG1 κ |