missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Bio-Techne NBP2-52717
Additional Details : Weight : 0.00970kg
Description
CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunoprecipitation.Specifications
CRISPR-Cas9 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunoprecipitation | |
1 mg/mL | |
Western Blot 1:500, Immunoprecipitation | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
CAS9 Synthetic peptide conjugated to KLH derived from within residues 1150-1200 of S. Pyogenes enzyme cas9 (Used CRISPR/Cas9 Genome Editing system ) | |
100 μL | |
Gene Editing, Gene Knockouts, Gene Therapy | |
Bacteria | |
IgG |