missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-52718
Additional Details : Weight : 0.00970kg
Description
CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in S. pyogenes samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CRISPR-Cas9 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
CRISPR-Cas9 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of Streptococcus pyogenes CRISPR-Cas9 protein. (Uniprot: Q99ZW2) | |
100 μg | |
Gene Editing, Gene Knockouts, Gene Therapy | |
S. pyogenes | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/ml | |
Western Blot 1:1000, ELISA 1:25000-1:30000, Immunocytochemistry/Immunofluorescence 1ug/mL, Immunofluorescence | |
Q99ZW2, Q99ZW2, Q99ZW2, Q99ZW2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |