missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crizotinib, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent inhibitor of c-MET and anaplastic lymphoma kinase (ALK) (cell IC50 values are 8.0 and 20 nM respectively). Selective for c-MET and ALK against >120 different kinases. Displays antitumor efficacy in multiple tumor models; inhibits c-MET-dependent proliferation, migration and invasion of human tumor cells in vitro. Orally bioavailable.
Specifications
Specifications
Assay Percent Range | >99% |
Quantity | 10 mg |
Formula Weight | 450.34 |
Chemical Name or Material | Crizotinib |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'