missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crocein Scarlet 7B, >70% dye content

Product Code. 15488409
Click to view available options
Quantity:
1g
5g
25g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15488409

Brand: Thermo Scientific Chemicals J66876.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6226-76-2 | C24H18N4Na2O7S2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6226-76-2
Molecular Formula C24H18N4Na2O7S2
Synonym Acid Red 71; C.I. 27165

Specifications

Color Red
Form Powder
Format Powder
Formula Weight 584.53
Melting Point 181-188°
Product Type Crocein Scarlet 7B
Purity >70% dye content
Quantity 25g
Solubility Soluble in water

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.