missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crocetin, free acid, >90%, MP Biomedicals™
Click to view available options
Quantity:
5 mg
Specifications
Color | Orange |
Assay Percent Range | >90% |
Quantity | 5 mg |
Formula Weight | 328.4 |
Physical Form | Powder |
Chemical Name or Material | Crocetin, free acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'