missing translation for 'onlineSavingsMsg'
Learn More

Cromolyn sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 15446739
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Mast cell stabilizer that prevents release of histamine from mast cells

Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.

Solubility
Soluble in water to 100mg/ml

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15826-37-6
Molecular Formula C23H14Na2O11
Molecular Weight (g/mol) 512.33
MDL Number MFCD00057744
InChI Key VLARUOGDXDTHEH-UHFFFAOYSA-L
Synonym cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal
PubChem CID 27503
ChEBI CHEBI:128458
SMILES [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O

Specifications

Color White to Yellow
Quantity 5 g
Merck Index 14,2590
Solubility Information Soluble in water to 100mg/ml
IUPAC Name disodium;5-[3-(2-carboxylato-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
Formula Weight 512.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Cromolyn sodium salt
compliance-icons
Product Identifier
  • Cromolyn sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Cromolyn sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.