missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Cross Linked Dextran Matrix Cytodex 3
£181.00 - £3645.00
Specifications
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area. |
---|---|
Product Type | Cytodex 3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329981
|
Cytiva
17-0485-01 |
10 g |
£181.00
Each |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11399971
|
Cytiva
17-0485-02 |
100 g |
£993.00
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11309981
|
Cytiva
17-0485-03 |
500 g |
£3645.00
500g |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cytodex 3, formed by chemically coupling a thin layer of denatured collagen to the cross-linked dextran matrix, is the microcarrier of choice for cells that may be difficult to culture in vitro. Microcarrier culture is a technique which makes possible the practical high yield culture of anchorage-dependent cells. Cytodex has been specifically developed for the culture of a wide range of animal cells in culture volumes ranging from a few milliliters to more than six thousand liters. Using Cytodex in simple suspension culture systems provides yields of several million cells per milliliter. Formed by substituting a cross-linked dextran matrix with positively charged DEAE groups distributed throughout the matrix, is a general purpose microcarrier.- Designed for culture of primary cells or those with an epithelioid morphology, especially cells with low plating-efficiency or differentiated or sensitive cell-types which are normally difficult to grow, e.g., hepatocytes, endocrine cells.
- Contains a layer of acid-denatured porcine collagen (gelatin) on the surface of the microporous matrix.
- Designed to yield harvested cells of the highest viability.
- Transparent for easy microscopic examination of attached cells.
Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area. | |
Cytodex 3 |