missing translation for 'onlineSavingsMsg'
Learn More

Crotonaldehyde, SPEX CertiPrep™

Product Code. 15226316
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Crotonaldehyde-(predom. trans)123-73-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 123-73-9, 7732-18-5
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C4H6O
MDL Number MFCD00007003
InChI Key MLUCVPSAIODCQM-NSCUHMNNSA-N
SMILES C\C=C\C=O
IUPAC Name (2E)-but-2-enal
Molecular Weight (g/mol) 70.09
PubChem CID 447466
ChEBI CHEBI:41607
Solvent or Matrix H2O
Chemical Name or Material Crotonaldehyde
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'