Learn More
Crotonyl chloride, tech. 90%, Thermo Scientific Chemicals
£16.60 - £214.00
Chemical Identifiers
CAS | 625-35-4 |
---|---|
Molecular Formula | C4H5ClO |
Molecular Weight (g/mol) | 104.533 |
MDL Number | MFCD00000734 |
InChI Key | RJUIDDKTATZJFE-NSCUHMNNSA-N |
Synonym | crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride |
PubChem CID | 643131 |
IUPAC Name | (E)-but-2-enoyl chloride |
SMILES | CC=CC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424714
|
Thermo Scientific Alfa Aesar
A13710.14 |
25 g |
£16.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434714
|
Thermo Scientific Alfa Aesar
A13710.22 |
100 g |
£54.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444714
|
Thermo Scientific Alfa Aesar
A13710.36 |
500 g |
£214.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Crotonyl chloride is used in agrochemical, pharmaceutical and dyestuff field. Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCrotonyl chloride is used in agrochemical, pharmaceutical and dyestuff field. Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store at 4°C. Store away from heat and moisture. Incompatible with
Chemical Identifiers
625-35-4 | |
104.533 | |
RJUIDDKTATZJFE-NSCUHMNNSA-N | |
643131 | |
CC=CC(=O)Cl |
C4H5ClO | |
MFCD00000734 | |
crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride | |
(E)-but-2-enoyl chloride |
Specifications
625-35-4 | |
121°C to 123°C | |
Acrid | |
1.46 | |
UN2920 | |
Moisture sensitive | |
Reacts with water. | |
CC=CC(=O)Cl | |
104.533 | |
104.54 | |
remainder mainly cis-isomer |
1.091 | |
35°C (95°F) | |
C4H5ClO | |
MFCD00000734 | |
506118 | |
crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride | |
RJUIDDKTATZJFE-NSCUHMNNSA-N | |
(E)-but-2-enoyl chloride | |
643131 | |
≥90% | |
Crotonyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 210-889-9
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only