missing translation for 'onlineSavingsMsg'
Learn More

Crotyl chloride, 70%, tech., Thermo Scientific Chemicals

£58.20 - £201.80

Specifications

Chemical Name or Material Crotyl chloride
Name Note tech., 70%
CAS 591-97-9, 563-52-0, 1190-22-3
Melting Point -65.0°C
Color Colorless
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10417560
View Documents
Thermo Scientific Acros
154801000
100 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10796551
View Documents
Thermo Scientific Acros
154805000
500 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Crotyl chloride
591-97-9, 563-52-0, 1190-22-3
Colorless
80.0°C to 85.0°C
Authentic
Glass bottle
CH3CH=CHCH2Cl
MFCD00000987
15, 2130
1-Chloro-2-butene
90.55
90.55
Technical
tech., 70%
-65.0°C
0.9200g/mL
−15°C
65% min. (GC)
C4H7Cl
1.4300 to 1.4360
01, II, 176
0.92
Solubility in water:+/- 14g/L (20°C)
0.46 mPa.s (20°C)
70%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 209-739-5

missing translation for 'rtecsNumber' : EM4264000

missing translation for 'tsca' : TSCA

Product Identifier
  • Crotyl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only