Learn More
CRP Recombinant Polyclonal Antibody (5HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710269
Description
This antibody is predicted to react with non-human primate, rat and rabbit based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
CRP Recombinant Polyclonal antibody specifically detects CRP in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CRP | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Aa1249; Ab1-341; Ab2-196; Ac1-114; Ac1262; Ac2-069; acute phase protein; CRP; AI255847; Ba2-693; c reactive; C-reactive preprotein; C-reactive protein; C-reactive protein member of the pentraxin family; C-reactive protein(1-205); C-reactive protein, member of the pentraxin family; C-reactive protein, pentraxin-related; C-reactive protein, petaxin related; CRP; CRP1; CSRP; CYRP; D1S181E; DKFZp686M148; DKFZp686M149; MGC149895; MGC88244; pentraxin 1; PTX1; RP11-419N10.4 | |
Recombinant protein corresponding to amino acids 119–224 of human CRP | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
5HCLC | |
Unconjugated | |
P02741, P14847 | |
Rabbit | |
Protein A | |
RUO | |
12944, 1401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |