missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Crucible Tongs

Bochem™ Crucible Tongs are rugged type Stainless Steel tongs.

Brand:  Bochem™ 2146

Additional Details : Weight : 0.25000kg

Product Code. 10140241

  • £47.95 / Each
Estimated Shipment: 11-10-2024
to see stock.
Add to basket

Description

Description

  • Stainless Steel
  • Rugged type
Specifications

Specifications

Max diameter: 400mm; Min diameter: 8mm
18/10 E-Poli
270 mm
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers