Learn More
Bochem™ Crucible Tongs
Biochem™ Crucible Tongs - Scissor-like tools with concave pincers that allow the user to grasp a hot crucible, flask, evaporating dish or small beaker.
Brand:  Bochem™ 2141
Additional Details : Weight : 0.25000kg
Product Code. 10376141
  • £17.90 / Each
Estimated Shipment: 04-10-2024
to see stock.
Add to basket
Description
Description
  • Crucible handling
  • Rugged type
  • Made from electrolytically polished stainless steel.
Specifications
Specifications
Concave Pincers
Crucible Tongs
220 mm
Handling Hot Crucibles, Flasks, Evaporating Dishes, Small Flasks
Stainless Steel (18/10)