missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Crustacean Cardioactive Peptide
For cellular and molecular biology applications
£181.00 - £373.00
Specifications
Purification Method | HPLC |
---|---|
Common Name | CCAP |
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 957.10 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15457819
|
Thermo Scientific Chemicals
J66453.MA |
10 mg |
£373.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15447819
|
Thermo Scientific Chemicals
J66453.#0 |
5 mg | N/A | ||||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
HPLC | |
-30°C to -10°C | |
957.10 | |
RUO - research use only |
CCAP | |
Solid | |
Crustacean Cardioactive Peptide | |
Pro-Phe-Cys-Asn-Ala-Phe-Thr-Gly-Cys (Disulfide bridge: Cys3-Cys9) |
RUO – Research Use Only