Learn More
Welch™ CRVpro™ Starter Package
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
Brand:  Welch™ 330073
 View more versions of this product
Product Code. 15716809
  • £3380.00 / Each
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CRVpro Starter Package Includes:
    • Oil mist separator AKD 16
    • 2-way ball valve
    • Hose connector DN 16KF-10/8 mm
    • Vacuum hose (1.5 m)
    • Oil change set
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRVpro with Oil Mist Separator AKD 16, 2-Way Ball Valve, Hose Connector, Vacuum Hose (1.5 m), Oil Change Set
CRVpro 8
Rotary Vane Vacuum Pump
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch™ CRVpro™ Starter Package > Vacuum Pump; Model: CRVpro 8