missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Gives protection in ultra-cold environments down to -196°C
- Maximum thermal protection, flexibility and dexterity
- Inner lining wicks away moisture for added comfort during prolonged use
- CE Cat. II, EN 511, EN 388, EN 420
Specifications
Specifications
Certifications/Compliance (Europe) | CE Category II, EN 511, EN 388, EN 420 |
Color | Blue |
Product Type | Cryogenic Gloves |
Size | Small |
Style | Mid-Arm |
Temperature (Metric) Max | -196°C |
For Use With (Application) | Handling dry ice, low and ultra-low temperature freezers, closed cryogenic systems |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tempshield™ Cryo-Gloves™ >
Spot an opportunity for improvement?Share a Content Correction