missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tempshield™ Cryo-Gloves™
Click to view available options
Style:
Mid-Arm
Shoulder
Description
- Gives protection in ultra-cold environments down to -196°C
- Maximum thermal protection, flexibility and dexterity
- Inner lining wicks away moisture for added comfort during prolonged use
- CE Cat. II, EN 511, EN 388, EN 420
Specifications
Certifications/Compliance (Europe) | CE Category II, EN 511, EN 388, EN 420 |
Color | Blue |
Product Type | Cryogenic Gloves |
Size | Small |
Style | Shoulder |
Temperature (Metric) Max | -196°C |
For Use With (Application) | Handling dry ice, low and ultra-low temperature freezers, closed cryogenic systems |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'