Learn More
Tempshield™ Cryo-Gloves™ Blue Mid-Arm Length Cryogenic Gloves
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Brand: Tempshield™ MAXL
Additional Details : Weight : 0.22500kg
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Cryogenic Gloves | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
33.5 to 39.5 cm |
Mid-Arm Length | |
Specific | |
X-Large | |
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems | |
-320°F | |
13.3 to 15.5 in. |