Learn More
Tempshield™ Cryo-Gloves™ Blue Waterproof Elbow Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
£434.00 - £445.00
Specifications
Certifications/Compliance (Europe) | EN 420, EN 511 |
---|---|
Description | Elbow Length |
Color | Blue |
Hand | Specific |
Product Type | Waterproof Cryogenic Gloves |
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11853801
|
Tempshield™
EBMWP |
Medium |
£445.00
1 pair |
In Stock
Log in to see stock. |
|||||
11893811
|
Tempshield™
EBLWP |
Large |
£445.00
1 pair |
In Stock
Log in to see stock. |
|||||
11873881
|
Tempshield™
EBXLWP |
X-Large |
£434.00
1 pair |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
Description
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Waterproof Cryogenic Gloves | |
Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes | |
-320°F | |
17.3 to 19.8 in. |
Elbow Length | |
Specific | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
44 to 50 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.