missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Cryo Gloves - Wrist Length Product Code.: 15659316
In Stock
to see stock.
Add to basket

Thermo Scientific™ Cryo Gloves - Wrist Length

Product Code. 15659316 Shop All Thermo Scientific Products
Large
Click to view available options
Size:
Small
Medium
Large

Product Code. 15659316

Brand: Thermo Scientific™ 189575

In Stock
to see stock.
Edge
Add to basket
Add to basket

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY

Specifications

Description Wrist Length
Color Blue
Size Large
For Use With (Application) Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English) 12 in.
Length (Metric) 30.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Cryo Gloves - Wrist Length >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.