missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tempshield™ Cryo-Industrial Mid-Arm Fabric Gloves
Click to view available options
Size:
Medium
Large
X-Large
Description
- Protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
- Gloves are designed to provide a high level of thermal protection and abrasion resistance for heavy-duty applications
- Mid-arm length
- Made from state-of-the-art materials
- Designed to keep the user dry and to wick moisture away from your hands
- Can be worn in comfort for extended periods of time.
- Multi-layered construction allows for a maximum level of thermal protection, flexibility, and dexterity
Specifications
Specifications
Color | Blue |
Material | Fabric |
Powdered | Powder Free |
Product Type | Cryogenic Frosters Mid-arm Gloves |
Size | Medium |
Sterility | Non-sterile |
Length (Metric) | 349 to 395 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'