missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Cryo.s Round Bottom 4 mL Polypropylene Tube with External Thread Cap
Ideal for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.
Brand: Greiner Bio-One 127263
This item is not returnable.
View return policy
Description
- With or without ID writing area
- With Star foot base
- Sterile
- Non-pyrogenic and non-toxic
- Includes 100 white cap inserts
- Polyethylener case
- Dia. x Height: 12.4 x 83mm
Service Plan
No

Specifications
12.4 mm | |
Natural | |
Round (Starfoot) | |
External | |
83 mm | |
Sterile |
Cryogenic Storage Tube | |
4 mL | |
Polypropylene | |
ID Writing Area, 150 Inserts | |
Polyethylene case | |
Threaded Screw Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Cryo.s Round Bottom 4 mL Polypropylene Tube with External Thread Cap > w/External thread natural cap; With ID writing area
Spot an opportunity for improvement?Share a Content Correction