Learn More
Eppendorf™ CryoCube™ F740hi with Touchscreen Interface
GREENER_CHOICE
Known for their energy efficiency, reliability, and worry-free, long-term sample preservation. The polyurethane foam insulation (incl. vacuum insuation panels for CryoCube F740) combined with a highly efficient cooling system achieve and maintain ultra-low temperatures of -86°C.
Brand:  Eppendorf™ F740320022
20344.55 GBP valid until 2025-12-31
Use promo code "25041" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15823262
  • £21415.32 / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conventionally foamed, reinforced by vacuum insulation panels in the device body and door reinforced polyurethane insulation ensures efficient insulation
  • External door with handle on the left or right for optimum space utilization in the laboratory
  • Easy-to-read, flush mounted control panel at eye level and alarm function
  • The insulated and sealed interior doors minimize cold losses when accessing the samples
  • Equipped with either three or five compartments that can be equipped with Eppendorf racks or other well-known brands
  • Each shelf has louvers for rapid temperature adjustment
  • Ultra Freezers are suitable for storage and preservation of all types of biological materials / samples
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Air-cooled
Right Handle
Vacuum, Polyurethane Foam
3 Shelves with Right Handle
Upright
5 years
Key Lock
CO2 or LN2 Optional
50/60 Hz
315 kg
740 L
3
3
CE
Gray
F740hi
7-day Ink or Inkless (factory installed)
230 V 50 Hz
10,500 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products