Learn More
SPEX Certiprep™ Cryogenic Grinding Vial Pack
The Vial Set is designed for grinding tough samples such as bone and plant material at room temperature using the Geno/Grinder
Brand: SPEX Certiprep™ 2255
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- A complete vial consists of 2 end plugs, a polycarbonate cylinder and 2 stainless steel 1/2” (12.7 mm) grinding balls.
- Vials are sold in sets of 4.
- Up to 16 vials can be run simultaneously
- Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
Specifications
Use with the SPEX homogeniser 2260 CryoBlock | |
Polycarbonate | |
4 Set |