missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Cryogenic Vial Boxes
GREENER_CHOICE

Select the size of the box required and match the partition to your tube requirements. Heathrow Scientific™ Cryogenic Vial Boxes are constructed from cardboard with vent holes to allow air circulation.

Brand:  Heathrow Scientific™ HS2860B

 View more versions of this product

Product Code. 11394015

  • £68.95 / Pack of 12
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Store in LN2, or in mechanical freezers
  • Colors allow for easy identification and sorting purposes
  • Bottom vent holes to allow quicker circulation and removal of cold air, vapor or LN2
  • All box footprints are 133 x 133mm, select from two different heights
  • Partitions must be ordered separately
Specifications

Specifications

White
Square
1.5 to 2 mL Tubes
12/Pk.
5.2 in.
5.2 in.
3 in.
Cardboard
Cryo Box
Lid
133 x 133 75 mm
13.3 cm
13.3 cm
7.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers