Learn More
Fisherbrand™ Cryogenic Vial Gripper
The perfect grip for optimal precision.
Brand:  Fisherbrand™ 120908F
Product Code. 17807155
  • £58.35 / Pack of 5
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear
  • The designated squeeze target ensures maximum precision and grip with every use
Specifications
Specifications
5.6 in.
Pack of 5
Non-sterile
14.2 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Cryogenic Vial Gripper > Fisherbrand Cryogenic/Vaccine Vial Grippers; Type: Cryogenic Vial; Color: Assorted; Pack: 5