Learn More
Thermo Scientific™ CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation needs.
£28310.04 - £32612.47
Especificaciones
Type | Controlled-rate freezer |
---|---|
Temperature Range | -180° to +50 °C |
Cabinet Material | Stainless Steel |
Exterior Finish | Powder-coated Steel |
Display | Touchscreen User Interface |
Código de producto | Marca | Model | Electrical Requirements | Capacity (Metric) | Precio | Cantidad y disponibilidad | |||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Model | Electrical Requirements | Capacity (Metric) | Precio | Cantidad y disponibilidad | |||
17171802
|
Thermo Scientific™
TSCM17PV |
General Purpose with Printer | 220-230 V, 50/60Hz | 17 L |
£29342.62
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||
17191802
|
Thermo Scientific™
TSCM34PV |
General Purpose with Printer | 220-230 V, 50/60Hz | 34 L |
£31149.65
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||
17111812
|
Thermo Scientific™
TSCM48PV |
General Purpose with Printer | 220-230 V, 50/60Hz | 48 L |
£32612.47
Each |
In Stock
Log in to see stock. |
|||
17181802
|
Thermo Scientific™
TSCM17XV |
General Purpose without Printer | 220-230 V, 50/60Hz | 17 L |
£28310.04
Each |
In Stock
Log in to see stock. |
|||
17101812
|
Thermo Scientific™
TSCM34XV |
General Purpose without Printer | 220-230 V, 50/60Hz | 34 L |
£30031.01
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||
17121812
|
Thermo Scientific™
TSCM48XV |
General Purpose without Printer | 220-230 V, 50/60Hz | 48 L |
£31579.89
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
Descripción
For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
User-friendly operation with enhanced data traceability
- Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
- Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
- Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
- USB data export of .pdf file run logs support 21 CFR part 11
- User security: 3 levels of user accessibility
- “Run last” feature allows the same profile to be run consecutively with the touch of a button
- PC interface software provides remote operation capabilities, run review, and custom profile creation
Real-time run monitoring for sample protection
- Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
- Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
- Remote alarm contacts
- Optional thermal printer
Reliable temperature performance for high throughput usage
- Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
- Preventative maintenance indicator for LN2 solenoid replacement limits downtime
- Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
- Compatible with VHP cleaning techniques
- Environmentally friendly insulating foam
- Type 304 stainless steel with exterior powder-coat finish
Warranty varies by region. Check with your local representative for more information
Especificaciones
Controlled-rate freezer | |
Stainless Steel | |
Touchscreen User Interface | |
Front-loading | |
Yes | |
Thermo Fisher Connect compatible |
-180° to +50 °C | |
Powder-coated Steel | |
Warranty varies by region. Contact your local sales representative for more information | |
USB data export | |
PC Interface Software Supported | |
12 A |