Learn More
MP Biomedicals™ Cryopres™ Dimethyl Sulfoxide (>99.9% USP DMSO for cryopreservation)
Commonly used in cell cryopreservation to prevent damage to the cell membrane during freezing.
Brand: MP Biomedicals™ 092780148
173.55 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : CAS Number : 67-68-5 Weight : 0.05500kg
Description
- Cryopres Dimethyl Sulfoxide (DMSO) USP, Ph. Eur. Sterile filtered
- Cryopres Dimethyl Sulfoxide is commonly used in cell cryopreservation, along with glycerol, to prevent damage to the cell membrane during freezing
- Complete Certificate of Analysis available each production lot, along with full formulation information
- Produced under the highest industry standards to ensure superior results
Specifications
15 to 30°C, desiccate, store under nitrogen | |
Sterile | |
0.999 |
Liquid | |
Cryopreservation | |
50 mL |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.