missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cryopreserved Hepatocyte Recovery Medium (CHRM™)

Product Code. 10292830
Click to view available options
Quantity:
50 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10292830

Brand: Gibco™ CM7000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Cryopreserved Hepatocyte Recovery Medium (CHRM)

Cryopreserved Hepatocyte Recovery Medium (CHRM®) is used for thawing Gibco cryopreserved human hepatocytes for optimal cell recovery and viability.

To ensure best results when using Gibco cryopreserved human hepatocytes, use a one step centrifugation process.

Related link
Learn more about our Hepatic Cell Culture reagents
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Cell Type Hepatocyte Cells
Classification FBS-supplemented
Form Liquid
Product Type Hepatocyte Supplement
Sterility Sterile
With Additives Phenol Red
Product Line Gibco
Quantity 50 mL
Remarks Proprietary formulation provided by AP Sciences
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.