Learn More
CellPath Cryospray Freezer Spray
Specially formulated to rapidly freeze fresh biopsy material removing the risk of morphological damage due to ice crystal formulation.
Brand:  CellPath KNA-0173-00A
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 12705308
  • £49.50 / 173g
Explore more special offers
Description
Description
  • CFC free
  • Directable jet enables precise control
  • Ideal for paraffin blocks and fresh tissue specimens
  • Manufactured from a mixture of environmentally friendly gases to give a good balance of cooling efficiency and cost
  • Supplied in a compact seamless aluminum can that is easy to handle, with a nozzle that ensures accurate direction of the freezing jet
Specifications
Specifications
175 mg
1 Pack
175 mg
Cryospray Freezer Spray
Ideal for paraffin blocks and fresh tissue specimens
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • CRYOSPRAY (FREEZER SPRAY)
Signal Word
  • Warning
Hazard Category
  • Flammable and non-flammable aerosols category 1
Hazard Statement
  • H229-Pressurised container: May burst if heated.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P251-Do not pierce or burn, even after use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P410-Protect from sunlight.
  • P412-Do not expose to temperatures exceeding 50°C/122°F.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1,1,1,2-Tetrafluoroethane