Learn More
Epredia™ Cryostat Cryostar™ Nx70 V Adds Vacutome Option
Brand: Epredia™ 957060
This item is not returnable.
View return policy
Description
With electronic height adjust, temp control, touch screen, motorised sectioning, large open chamber 230V, 50/60 Hz, 1000 x 820 mm to 1120 x 755 mm Epredia™ Shandon