missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ CryoStorage Vials, 3D SafeCode

Product Code. 16416879 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.5 mL
1.0 mL
1.5 mL
2.0 mL
4.0 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 480
Pack of 960
This item is not returnable. View return policy

Product Code. 16416879

Brand: Eppendorf™ 0030079442

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides a 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labeled vials are pre-racked and pre-capped (0.5 mL to 4.0 mL) for storage applications.

  • 3-level coding for safe sample identification
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -196 °C (gas phase of LN2) to +121 °C
  • Manufactured from high-quality polypropylene in class VIII cleanroom for highest sample purity
  • Pre-capped and pre-racked for comfortable handling
  • Additional information on vials available via Eppendorf website for documentation purposes
  • Direct import into Eppendorf eLABInventory and eLABJournal software for convenient handling
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Sterility Non-sterile
Volume (Metric) 4.0 mL
No. per Pack 48
RNase Free Yes
Dnase Free Yes
Barcode 2D SafeCode
Quantity 480 (10 Racks of 48 Vials)
Type Coded Cryobank Vial System
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.