Learn More
Epredia™ Cryotome™ Cryostat Cryocassettes, 25mm dia. Crosshatched
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Epredia™ Cryotome™ Cryostat Accessories.
Brand:  Epredia™ A78910078
Code : 50
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12067119
  • £18.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Description
Description
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on active Peltier
  • Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
Specifications
Specifications
Knife Guard
Knife Guard
FSE/FE/SME/E