Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing with Three Stop Configuration
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing with Three Stop Configuration handles virtually all inorganic chemicals found in the lab.
Brand: Saint Gobain Life Sciences 070535-17-ND
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Tubing | |
Yellow/Blue | |
1.52 mm | |
0.13 in. | |
32.8 ft. | |
0.03 in. | |
For sample and reagent dispensing, biological and chemical assays, and water and soil analysis | |
12 Pk. | |
Crystal Clear | |
Flexible |
FDA Compliant, NSF, 3-A, USP Class VI | |
0.06 in. | |
Tygon E-3607 | |
3.22 mm | |
10 m | |
0.85 mm | |
-50°C | |
1.52 x 3.22 mm | |
55 (Shore A) | |
74°C |