missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ LMT-55 Tubing without Configuration

Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing without Configuration handles virtually all inorganic chemicals found in the lab.

Brand:  Saint Gobain Life Sciences 070534-11L-ND

 View more versions of this product

Product Code. 15133226

  • £58.75 / 10m
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
  • Resistant to flex fatigue and abrasion
  • Less permeable than rubber tubing
  • For use with virtually all inorganic chemicals found in the lab
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FDA Compliant, NSF, 3-A, USP Class VI
0.04 in.
Tygon R3607
2.72 mm
10 m
0.85 mm
-58°F
1 Ea.
55 (Shore A)
165.2°F
Tubing
1.02 mm
0.11 in.
32.8 ft.
0.03 in.
For sample and reagent dispensing, biological and chemical assays, and water and soil analysis
-50°C
Crystal Clear
Flexible
74°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers