missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet, certified

£21.61 - £155.78

Specifications

Chemical Name or Material Crystal Violet
CAS 548-62-9, 90-94-8
Absorption Ratio 0.98 to 1.2 (P-15/P+15)
Dye Content 88% min.
Molecular Formula C25H30ClN3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10626621
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
405830250
25 g
£21.61
25g
Please to purchase this item. Need a web account? Register with us today!
10390184
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
405831000
100 g
£49.75
100g
Please to purchase this item. Need a web account? Register with us today!
13488548
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
405835000
500 g
£155.78
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Crystal Violet
0.98 to 1.2 (P-15/P+15)
C25H30ClN3
13, 756
Basic Violet 3, C.I. 42555, Gentian Violet
Authentic
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
Green
548-62-9, 90-94-8
88% min.
MFCD00011750
15, 4430
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Certified
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
May cause cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • Crystal Violet

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P273-Avoid release to the environment.

Supplemental information
  • MIXTURE LIST-Contains : C.I. Basic violet 1,Michler's ketone

SDS
Documents

Documents

Certificates

RUO – Research Use Only